Ethyl-6-bromoimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate

046920-500MG 046920-1G
101877-642EA 181.54 USD
101877-642 101877-644
Ethyl-6-bromoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉BrN₂O₂
MW: 269.10 g/mol
MDL Number: MFCD06659034
CAS Number: 67625-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR