5-Bromo-1-(4-methylbenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-1,6-dihydro-1-(4-methylbenzyl)-6-oxopyridine-3-carboxaldehyde

046908-500MG 046908-1G
101877-580EA 747.69 USD
101877-580 101877-582
5-Bromo-1-(4-methylbenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
5-Bromo-1-(4-methylbenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₂BrNO₂
MW: 306.17 g/mol
MDL Number: MFCD09817447
CAS Number: 952183-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR