tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

046875-1G 046875-5G
101877-404EA 143.08 USD
101877-404 101877-406
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate

Hazard: Irritant-Harmful/Refrigerate

Formula: C₁₁H₂₃N₃O₂
Melting Pt: 36…41 °C
Flash Pt: >198 °C
MDL Number: MFCD02683049
CAS Number: 192130-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR