Ethyl-2-bromo-3-oxo-3-phenylpropanoate ≥95%

Supplier: Matrix Scientific

046850-500MG 046850-1G 046850-5G
101877-266EA 127.69 USD
101877-266 101877-268 101877-270
Ethyl-2-bromo-3-oxo-3-phenylpropanoate ≥95%
Ethyl-2-bromo-3-oxo-3-phenylpropanoate

Hazard: Irritant

Formula: C₁₁H₁₁BrO₃
MW: 271.12 g/mol
MDL Number: MFCD08444026
CAS Number: 55919-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR