1-(6-Methyl-5-nitro-1H-indazol-1-yl)-1-ethanone ≥95%

Supplier: Matrix Scientific

046839-500MG 046839-1G
101877-206EA 556.92 USD
101877-206 101877-208
1-(6-Methyl-5-nitro-1H-indazol-1-yl)-1-ethanone ≥95%
1-(6-Methyl-5-nitro-1H-indazol-1-yl)-1-ethanone

Hazard: Irritant

Formula: C₁₀H₉N₃O₃
MDL Number: MFCD12025920
CAS Number: 81115-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR