3-Bromo-4-nitro-1H-indazole ≥95%
Supplier: Matrix Scientific
046838-500MG
046838-1G
046838-5G
101877-200EA
473.85
USD
101877-200
101877-202
101877-204
3-Bromo-4-nitro-1H-indazole ≥95%
3-Bromo-4-nitro-1H-indazole
Hazard: Irritant
Formula:
C₇H₄BrN₃O₂ |
MDL Number:
MFCD07781572 CAS Number: 74209-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...