5-Bromo-2-(methylsulfonyl)pyrimidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

046837-500MG 046837-1G
101877-196EA 375.38 USD
101877-196 101877-198
5-Bromo-2-(methylsulfonyl)pyrimidine-4-carboxylic acid ≥95%
5-Bromo-2-(methylsulfonyl)pyrimidine-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₅BrN₂O₄S
MDL Number: MFCD06742786
CAS Number: 30321-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR