Methyl-5-bromo-2-(methylsulfonyl)-4-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

046836-500MG 046836-1G
101877-192EA 376.92 USD
101877-192 101877-194
Methyl-5-bromo-2-(methylsulfonyl)-4-pyrimidinecarboxylate ≥95%
Methyl-5-bromo-2-(methylsulfonyl)-4-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₇H₇BrN₂O₄S
MW: 295.12 g/mol
MDL Number: MFCD11100780
CAS Number: 1060795-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR