[2,3-Dichloro-5-(trifluoromethyl)-4-pyridinyl]methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
2,3-Dichloro-4-(hydroxymethyl)-5-(trifluoromethyl)pyridine
046818-500MG
046818-1G
101877-132EA
1296.92
USD
101877-132
101877-134
[2,3-Dichloro-5-(trifluoromethyl)-4-pyridinyl]methanol ≥95%
[2,3-Dichloro-5-(trifluoromethyl)-4-pyridinyl]methanol
Hazard: Irritant
Formula:
C₇H₄Cl₂F₃NO MW: 246.02 g/mol |
MDL Number:
MFCD12025911 CAS Number: 1160474-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...