5-Nitro-2-[4-(trifluoromethyl)piperidino]benzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
046785-500MG
046785-1G
101876-998EA
375.38
USD
101876-998
101877-000
5-Nitro-2-[4-(trifluoromethyl)piperidino]benzenecarbaldehyde ≥95%
5-Nitro-2-[4-(trifluoromethyl)piperidino]benzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₃F₃N₂O₃ MW: 302.26 g/mol |
MDL Number:
MFCD10758093 CAS Number: 1033463-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...