6-[4-(Trifluoromethyl)piperidino]nicotinic acid ≥95%

Supplier: Matrix Scientific

046783-500MG 046783-1G
101876-992EA 375.38 USD
101876-992 101876-994
6-[4-(Trifluoromethyl)piperidino]nicotinic acid ≥95%
6-[4-(Trifluoromethyl)piperidino]nicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₃F₃N₂O₂
MDL Number: MFCD10758091
CAS Number: 1033463-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR