Ethyl-4-cyano-3-fluorobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-cyano-3-fluorobenzoate

046733-1G 046733-5G
101876-780EA 89.23 USD
101876-780 102833-146
Ethyl-4-cyano-3-fluorobenzenecarboxylate ≥95%
Ethyl-4-cyano-3-fluorobenzenecarboxylate

Hazard: Irritant

Formula: C₁₀H₈FNO₂
MDL Number: MFCD09607944
CAS Number: 952183-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR