3-Acetyl-2-chloropyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-acetyl-2-chloropyridine, 1-(2-Chloro-3-pyridinyl)-1-ethanone, 1-(2-Chloropyridin-3-yl)ethanone

046731-1G 046731-5G
101876-770EA 21.54 USD
101876-770 101876-772
3-Acetyl-2-chloropyridine ≥95%
3-Acetyl-2-chloropyridine

Hazard: Irritant

Formula: C₇H₆ClNO
MW: 155.5816 g/mol
MDL Number: MFCD03840751
CAS Number: 55676-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR