tert-Butyl-N-[(phenylsulfonyl)(2-thienyl)methyl]carbamate ≥95%

Supplier: Matrix Scientific

046730-500MG 046730-1G 046730-5G
101876-764EA 181.54 USD
101876-764 101876-766 101876-768
tert-Butyl-N-[(phenylsulfonyl)(2-thienyl)methyl]carbamate ≥95%
tert-Butyl-N-[(phenylsulfonyl)(2-thienyl)methyl]carbamate

Hazard: Irritant

Formula: C₁₆H₁₉NO₄S₂
MDL Number: MFCD09817433
CAS Number: 479423-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR