4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

046694-500MG 046694-1G 046694-5G
101876-580EA 318.46 USD
101876-580 101876-582 101876-584
4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride ≥95%
4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride

Hazard: Irritant

Formula: C₁₂H₇ClF₃NO₃S
MDL Number: MFCD07774162
CAS Number: 874838-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR