tert-Butyl-3-(4-chlorobenzyl)-3-formyltetrahydro-1(2H)-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(4-Chlorobenzyl)-3-formylpiperidine, N-BOC protected
046674-500MG
046674-1G
101876-500EA
1109.23
USD
101876-500
101876-502
tert-Butyl-3-(4-chlorobenzyl)-3-formyltetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-3-(4-chlorobenzyl)-3-formyltetrahydro-1(2H)-pyridinecarboxylate
Hazard: Irritant
Formula:
C₁₈H₂₄ClNO₃ MW: 337.85 g/mol |
MDL Number:
MFCD09607940 CAS Number: 952183-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...