Ethyl-2-[cyclohexyl(methyl)amino]nicotinate ≥95%

Supplier: Matrix Scientific

046658-500MG 046658-1G 046658-5G
101876-440EA 181.54 USD
101876-440 101876-442 101876-444
Ethyl-2-[cyclohexyl(methyl)amino]nicotinate ≥95%
Ethyl-2-[cyclohexyl(methyl)amino]nicotinate

Hazard: Irritant

Formula: C₁₅H₂₂N₂O₂
MDL Number: MFCD12025888
CAS Number: 1147979-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR