Boc-(R)-3-amino-4-(2-thienyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(thiophen-2-yl)butanoic acid

042206-250MG 042206-1G
101854-890EA 412.31 USD
101854-890 101854-892
Boc-(R)-3-amino-4-(2-thienyl)butyric acid
Boc-(R)-3-amino-4-(2-thienyl)butyric acid

Hazard: Irritant

Formula: C₁₃H₁₉NO₄S
MW: 285.36 g/mol
MDL Number: MFCD01860984
CAS Number: 269726-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR