Boc-(R)-3-amino-4-(2-naphthyl)butyric acid

Supplier: Matrix Scientific

042205-250MG 042205-1G
101854-886EA 275.38 USD
101854-886 101854-888
Boc-(R)-3-amino-4-(2-naphthyl)butyric acid
Boc-(R)-3-amino-4-(2-naphthyl)butyric acid

Hazard: Irritant

Formula: C₁₉H₂₃NO₄
MW: 329.4 g/mol
MDL Number: MFCD01076283
CAS Number: 219297-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR