Boc-(R)-3-amino-4-(2-furyl)butyric acid

Supplier: Matrix Scientific

042203-250MG 042203-1G
101854-878EA 498.46 USD
101854-878 101854-880
Boc-(R)-3-amino-4-(2-furyl)butyric acid
Boc-(R)-3-amino-4-(2-furyl)butyric acid

Hazard: Irritant

Formula: C₁₃H₁₉NO₅
MW: 269.3 g/mol
MDL Number: MFCD01860993
CAS Number: 270596-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR