Boc-(R)-3-amino-4-(2-fluorophenyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(R)-3-((tert-Butoxycarbonyl)amino)-4-(2-fluorophenyl)butanoic acid
042202-250MG
042202-1G
101854-874EA
275.38
USD
101854-874
101854-876
Boc-(R)-3-amino-4-(2-fluorophenyl)butyric acid
Boc-(R)-3-amino-4-(2-fluorophenyl)butyric acid
Hazard: Irritant
Formula:
C₁₅H₂₀FNO₄ MW: 297.33 g/mol |
MDL Number:
MFCD01076278 CAS Number: 218608-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...