Boc-(R)-3-amino-4-(2-chlorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-N-Boc-3-Amino-4-(2-chlorophenyl)butanoic acid

042200-250MG 042200-1G
101854-866EA 149.23 USD
101854-866 101854-868
Boc-(R)-3-amino-4-(2-chlorophenyl)butyric acid
Boc-(R)-3-amino-4-(2-chlorophenyl)butyric acid

Hazard: Irritant

Formula: C₁₅H₂₀ClNO₄
MW: 313.78 g/mol
MDL Number: MFCD01076274
CAS Number: 218608-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR