Boc-(R)-3-amino-4-(2,4-dichlorophenyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(R)-3-((tert-Butoxycarbonyl)amino)-4-(2,4-dichlorophenyl)butanoic acid
042199-250MG
042199-1G
101854-862EA
349.23
USD
101854-862
101854-864
Boc-(R)-3-amino-4-(2,4-dichlorophenyl)butyric acid
Boc-(R)-3-amino-4-(2,4-dichlorophenyl)butyric acid
Hazard: Irritant
Formula:
C₁₅H₁₉Cl₂NO₄ MW: 348.23 g/mol |
MDL Number:
MFCD01860946 CAS Number: 269396-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...