Boc-(R)-3-amino-4-(1-naphthyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(naphthalen-1-yl)butanoic acid, Boc-D-β-HoAla(1-Naphthyl)-OH

042198-250MG 042198-1G
101854-858EA 138.46 USD
101854-858 101854-860
Boc-(R)-3-amino-4-(1-naphthyl)butyric acid
Boc-(R)-3-amino-4-(1-naphthyl)butyric acid

Hazard: Irritant

Formula: C₁₉H₂₃NO₄
MW: 329.4 g/mol
MDL Number: MFCD01076281
CAS Number: 190190-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR