Boc-(R)-1,2,3,4-tetrahydroisoquinoline-3-acetic acid
Supplier: Matrix Scientific
Synonyms:
(R)-2-(2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinolin-3-yl)acetic acid
042197-250MG
042197-1G
101854-854EA
373.85
USD
101854-854
101854-856
Boc-(R)-1,2,3,4-tetrahydroisoquinoline-3-acetic acid
Boc-(R)-1,2,3,4-tetrahydroisoquinoline-3-acetic acid
Hazard: Irritant
Formula:
C₁₆H₂₁NO₄ MW: 291.35 g/mol |
MDL Number:
MFCD01860933 CAS Number: 332064-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...