Fmoc-4-(3-aminophenyl)butanoic acid
Supplier: Matrix Scientific
042192-100MG
042192-500MG
101854-834EA
275.38
USD
101854-834
101854-836
Fmoc-4-(3-aminophenyl)butanoic acid
Fmoc-4-(3-aminophenyl)butanoic acid
Hazard: Irritant
Formula:
C₂₅H₂₃NO₄ |
MDL Number:
MFCD02682316 CAS Number: 867382-45-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...