Fmoc-(4-aminophenyl)acetic acid
Supplier: Matrix Scientific
042186-1G
042186-5G
101854-810EA
386.15
USD
101854-810
101854-812
Fmoc-(4-aminophenyl)acetic acid
Fmoc-(4-aminophenyl)acetic acid
Hazard: Irritant
Formula:
C₂₃H₁₉NO₄ |
MDL Number:
MFCD01861339 CAS Number: 173690-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...