Fmoc-(4-aminomethylphenyl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(4-(((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)methyl)phenyl)acetic acid, Fmoc-4-aminomethyl-phenylacetic acid

042185-1G 042185-5G
101854-806EA 386.15 USD
101854-806 101854-808
Fmoc-(4-aminomethylphenyl)acetic acid
Fmoc-(4-aminomethylphenyl)acetic acid

Hazard: Irritant

Formula: C₂₄H₂₁NO₄
MW: 387.44 g/mol
MDL Number: MFCD01321022
CAS Number: 176504-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR