Fmoc-(3-aminomethylphenyl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(3-(((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)methyl)phenyl)acetic acid

042183-1G 042183-5G
101854-798EA 461.54 USD
101854-798 101854-800
Fmoc-(3-aminomethylphenyl)acetic acid
Fmoc-(3-aminomethylphenyl)acetic acid

Hazard: Irritant

Formula: C₂₄H₂₁NO₄
MW: 387.44 g/mol
MDL Number: MFCD06656460
CAS Number: 631915-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR