Di-boc-2-amino-3-(2-aminobenzoyl) propionic acid
Supplier: Matrix Scientific
042178-500MG
042178-1G
101854-778EA
536.92
USD
101854-778
101854-780
Di-boc-2-amino-3-(2-aminobenzoyl) propionic acid
Di-boc-2-amino-3-(2-aminobenzoyl) propionic acid
Hazard: Irritant
Formula:
C₂₀H₂₈N₂O₇ |
MDL Number:
MFCD04974541 CAS Number: 1219434-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...