Boc-4-aminohippuric acid
Supplier: Matrix Scientific
042176-1G
042176-5G
101854-770EA
326.15
USD
101854-770
101854-772
Boc-4-aminohippuric acid
Boc-4-aminohippuric acid
Hazard: Irritant
Formula:
C₁₄H₁₈N₂O₅ |
MDL Number:
MFCD02682307 CAS Number: 854621-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...