Boc-4-(4-aminophenyl)butanoic acid

Supplier: Matrix Scientific

Synonyms: 4-(4-((tert-Butoxycarbonyl)amino)phenyl)butanoic acid, 4-(Boc-4-aminophenyl)butanoic acid

042174-1G 042174-5G
101854-762EA 498.46 USD
101854-762 101854-764
Boc-4-(4-aminophenyl)butanoic acid
Boc-4-(4-aminophenyl)butanoic acid

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
MDL Number: MFCD06656431
CAS Number: 105300-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR