Boc-4-(3-aminophenyl)butanoic acid

Supplier: Matrix Scientific

042173-100MG 042173-500MG
101854-758EA 275.38 USD
101854-758 101854-760
Boc-4-(3-aminophenyl)butanoic acid
Boc-4-(3-aminophenyl)butanoic acid

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD02682315
CAS Number: 149505-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR