Boc-4-(2-aminophenyl)butanoic acid

Supplier: Matrix Scientific

042172-100MG 042172-500MG
101854-754EA 275.38 USD
101854-754 101854-756
Boc-4-(2-aminophenyl)butanoic acid
Boc-4-(2-aminophenyl)butanoic acid

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD09750484
CAS Number: 214829-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR