3-(Boc-amino)-3-phenylpropionic acid
Supplier: Matrix Scientific
Synonyms:
Boc-DL-β-Phe-OH, 3-tert-Butoxycarbonylamino-3-phenyl-propionic acid
042171-1G
042171-5G
101854-750EA
275.38
USD
101854-750
101854-752
3-(Boc-amino)-3-phenylpropionic acid
3-(Boc-amino)-3-phenylpropionic acid
Hazard: Irritant
Formula:
C₁₄H₁₉NO₄ MW: 265.31 g/mol |
MDL Number:
MFCD02090695 CAS Number: 14676-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...