Boc-3-amino-2-naphthoic acid

Supplier: Matrix Scientific

042170-1G 042170-5G
101854-746EA 249.23 USD
101854-746 101854-748
Boc-3-amino-2-naphthoic acid
Boc-3-amino-2-naphthoic acid

Hazard: Irritant

Formula: C₁₆H₁₇NO₄
MW: 287.32 g/mol
MDL Number: MFCD02682309
CAS Number: 887242-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR