Boc-3-(4-aminophenyl)propionic acid

Supplier: Matrix Scientific

042169-1G 042169-5G
101854-742EA 598.46 USD
101854-742 101854-744
Boc-3-(4-aminophenyl)propionic acid
Boc-3-(4-aminophenyl)propionic acid

Hazard: Irritant

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
MDL Number: MFCD06656430
CAS Number: 149506-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR