Boc-(4-aminophenylthio)acetic acid

Supplier: Matrix Scientific

042168-1G 042168-5G
101854-738EA 498.46 USD
101854-738 101854-740
Boc-(4-aminophenylthio)acetic acid
Boc-(4-aminophenylthio)acetic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₄S
MDL Number: MFCD02682317
CAS Number: 447414-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR