Boc-(3-aminomethylphenyl)acetic acid

Supplier: Matrix Scientific

Synonyms: 3-[(Boc-amino)methyl]phenylacetic Acid

042165-1G 042165-5G
101854-726EA 260 USD
101854-726 101854-728
Boc-(3-aminomethylphenyl)acetic acid
Boc-(3-aminomethylphenyl)acetic acid

Hazard: Irritant

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
MDL Number: MFCD06656429
CAS Number: 71420-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR