Boc-(2R,3S)-2-amino-3-(phenylthio)butanoic acid
Supplier: Matrix Scientific
042164-100MG
042164-500MG
101854-722EA
598.46
USD
101854-722
101854-724
Boc-(2R,3S)-2-amino-3-(phenylthio)butanoic acid
Boc-(2R,3S)-2-amino-3-(phenylthio)butanoic acid
Hazard: Irritant
Formula:
C₁₅H₂₁NO₄S MW: 311.4 g/mol |
MDL Number:
MFCD04974544 CAS Number: 1217617-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...