Fmoc-L-1,2,3,4-tetrahydronorharman-3-carboxylicacid

Supplier: Matrix Scientific

Synonyms: (S)-2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid

042161-1G 042161-5G
101854-714EA 113.85 USD
101854-714 101854-716
Fmoc-L-1,2,3,4-tetrahydronorharman-3-carboxylicacid
Fmoc-L-1,2,3,4-tetrahydronorharman-3-carboxylicacid

Hazard: Irritant

Formula: C₂₇H₂₂N₂O₄
MW: 438.49 g/mol
MDL Number: MFCD00673786
CAS Number: 204322-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR