Fmoc-DL-7-azatryptophan

Supplier: Matrix Scientific

042160-1G 042160-5G
101854-710EA 958.46 USD
101854-710 101854-712
Fmoc-DL-7-azatryptophan
Fmoc-DL-7-azatryptophan

Hazard: Irritant

Formula: C₂₅H₂₁N₃O₄
MW: 427.46 g/mol
MDL Number: MFCD02682351
CAS Number: 1219143-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR