Fmoc-DL-6-methoxy-1,2,3,4-tetrahydronorharman-1-carboxylic acid

Supplier: Matrix Scientific

042159-250MG 042159-1G
101854-706EA 373.85 USD
101854-706 101854-708
Fmoc-DL-6-methoxy-1,2,3,4-tetrahydronorharman-1-carboxylic acid
Fmoc-DL-6-methoxy-1,2,3,4-tetrahydronorharman-1-carboxylic acid

Hazard: Irritant

Formula: C₂₈H₂₄N₂O₅
MDL Number: MFCD02682414
CAS Number: 1219416-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR