Fmoc-D-1,2,3,4-tetrahydro-norharman-3-carboxylicacid

Supplier: Matrix Scientific

Synonyms: (R)-2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid

042158-1G 042158-5G
101854-702EA 113.85 USD
101854-702 101854-704
Fmoc-D-1,2,3,4-tetrahydro-norharman-3-carboxylicacid
Fmoc-D-1,2,3,4-tetrahydro-norharman-3-carboxylicacid

Hazard: Irritant

Formula: C₂₇H₂₂N₂O₄
MW: 438.49 g/mol
MDL Number: MFCD01861321
CAS Number: 268731-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR