Fmoc-α-methyl-DL-tryptophan

Supplier: Matrix Scientific

042157-100MG 042157-1G
101854-698EA 249.23 USD
101854-698 101854-700
Fmoc-α-methyl-DL-tryptophan
Fmoc-alpha -methyl-DL-tryptophan

Hazard: Irritant

Formula: C₂₇H₂₄N₂O₄
MW: 440.5 g/mol
MDL Number: MFCD02682650
CAS Number: 114779-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR