Fmoc-7-bromo-DL-tryptophan
Supplier: Matrix Scientific
042155-100MG
042155-1G
101854-690EA
326.15
USD
101854-690
101854-692
Fmoc-7-bromo-DL-tryptophan
Fmoc-7-bromo-DL-tryptophan
Hazard: Irritant
Formula:
C₂₆H₂₁BrN₂O₄ |
MDL Number:
MFCD09750491 CAS Number: 1219370-30-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...