Fmoc-7-benzyloxy-DL-tryptophan

Supplier: Matrix Scientific

042154-10MG 042154-100MG
101854-686EA 598.46 USD
101854-686 101854-688
Fmoc-7-benzyloxy-DL-tryptophan
Fmoc-7-benzyloxy-DL-tryptophan

Hazard: Irritant

Formula: C₃₃H₂₈N₂O₅
MW: 532.6 g/mol
MDL Number: MFCD02682355
CAS Number: 1219174-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR