Fmoc-6-methyl-DL-tryptophan
Supplier: Matrix Scientific
042153-100MG
042153-1G
101854-682EA
349.23
USD
101854-682
101854-684
Fmoc-6-methyl-DL-tryptophan
Fmoc-6-methyl-DL-tryptophan
Hazard: Irritant
Formula:
C₂₇H₂₄N₂O₄ |
MDL Number:
MFCD02682377 CAS Number: 1219150-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...