Fmoc-6-chloro-D-tryptophan

Supplier: Matrix Scientific

042151-100MG 042151-500MG
101854-674EA 1643.08 USD
101854-674 101854-676
Fmoc-6-chloro-D-tryptophan
Fmoc-6-chloro-D-tryptophan

Hazard: Irritant

Formula: C₂₆H₂₁ClN₂O₄
MDL Number: MFCD09750494
CAS Number: 925916-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR