Fmoc-6-chloro-DL-tryptophan

Supplier: Matrix Scientific

042150-100MG 042150-1G
101854-670EA 363.08 USD
101854-670 101854-672
Fmoc-6-chloro-DL-tryptophan
Fmoc-6-chloro-DL-tryptophan

Hazard: Irritant

Formula: C₂₆H₂₁ClN₂O₄
MW: 460.92 g/mol
MDL Number: MFCD09750496
CAS Number: 1219168-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR